missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-9R Antibody (33423), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB290B
This item is not returnable.
View return policy
Description
IL-9R Monoclonal antibody specifically detects IL-9R in Human samples. It is validated for Flow Cytometry, Neutralization, CyTOF
Specifications
| IL-9R | |
| Monoclonal | |
| Biotin | |
| PBS | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Neutralization, CyTOF | |
| 33423 | |
| Flow Cytometry, Neutralization, CyTOF-ready | |
| CD129, CD129 antigen, IL-9 receptor, IL-9R, interleukin 9 receptor, interleukin-9 receptor | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-9 R | |
| 0.5 mL | |
| Cytokine Research | |
| 3581 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction