missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL10RB Polyclonal Antibody
GREENER_CHOICE

Product Code. 16689864
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16689864 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16689864 Supplier Invitrogen™ Supplier No. PA5103417

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total IL10RB.

Natural killer (NK) cells in teleosts and their evolutionary homologue are a subpopulation of lymphocytes with properties that distinguish them from either B- or T-cells. NK cells are important effectors of innate immunity where they release cytokines, which in turn up-regulate other immunological functions. Monoclonal antibodies have been used to identify different surface antigens present on NK cells. These surface antigens have not only been used to identify NK cells, but also their functionally distinct subsets. The Cluster of Differentiation (CD) nomenclature was established to standardize the naming of NK cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL10RB
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene IL10RB
Gene Accession No. Q08334
Gene Alias 6620401D04Rik; AI528744; CDW210B; CRF2-4; CRFB4; cytokine receptor class-II CRF2-4; cytokine receptor class-II member 4; Cytokine receptor family 2 member 4; cytokine receptor family II, member 4; D16H21S58; D21S58; D21S58h; D21S66; IL-10 receptor subunit beta; IL-10R subunit 2; IL-10R subunit beta; Il10r2; IL-10R2; IL10RB; IL-10RB; inte; interleukin 10 receptor 2; interleukin 10 receptor subunit beta; interleukin 10 receptor, beta; interleukin-10 receptor subunit 2; Interleukin-10 receptor subunit beta; RGD1560373
Gene Symbols IL10RB
Host Species Rabbit
Immunogen A synthesized peptide derived from human IL10RB(Accession Q08334), corresponding to amino acid residues S286-S325.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3588
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.