missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL11RA, Mouse anti-Human, Clone: 3D4-G4, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00003590-M02.100ug
This item is not returnable.
View return policy
Description
Sequence: MSSSCSGLSRVLVAVATALVSASSPCPQAWGPPGVQYGQPGRSVKLCCPGVTAGDPVSWFRDGEPKLLQGPDSGLGHELVLAQADSTDEGTYICQTLDGALGGTVTLQLGYPPARPVVSCQAADYENFSCTWSPSQISGLPTRYLTSYRKKTVLGADSQRRSPSTGPWPCPQDPLGAARCVVHGAEFWSQYRINVTEVNPLGASTRLLDVSLQSILRPDPPQGLRVESVPGYPRRLRASWTYPASWPCQPHFLLKFRLQYRPAQHPAWSTVEPAGLEEVITDAVAGLPHAVRVSARDFLDAGTWSTWSPEAWGTPSTGTIPKEIPAWGQLHTQPEVEPQVDSPAPPRPSLQPHPRLLDHRDSVEQVAVLASLGTLSFLGLVAGALALGLWLRLRRGGKDGSPKPGFLASVIPVDRRPGAPNLSpecifications
| IL11RA | |
| Monoclonal | |
| Unconjugated | |
| interleukin 11 receptor, alpha | |
| MGC2146 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 3D4-G4 | |
| In 1x PBS, pH 7.4 | |
| BC003110 | |
| IL11RA | |
| IL11RA (AAH03110, 1 a.a. ∼ 422 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 3590 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction