missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ IL12B Antibody (ebdarokimab) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
€ 213.00 - € 920.00
Specifications
| Antigen | IL12B |
|---|---|
| Clone | ebdarokimab |
| Applications | Flow Cytometry, ELISA, Functional Assay |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30047179
|
Novus Biologicals™
NBP3-28551-1mg |
1 mg |
€ 920.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30046854
|
Novus Biologicals™
NBP3-28551-50ug |
50 μg |
€ 213.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30046363
|
Novus Biologicals™
NBP3-28551-100ug |
100 μg |
€ 326.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL12B Monoclonal antibody specifically detects IL12B in Human samples. It is validated for Flow Cytometry,ELISA,Functional AssaySpecifications
| IL12B | |
| Flow Cytometry, ELISA, Functional Assay | |
| Unconjugated | |
| Human | |
| Cell Cycle and Replication, Cytokine Research, Immunology, Innate Immunity | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| CLMF, CLMF p40, CLMF2, Cytotoxic lymphocyte maturation factor 40 kDa subunit, IL-12 subunit p40, IL-12BNK cell stimulatory factor chain 2, interleukin 12, p40, interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocytematuration factor 2, p40), interleukin-12 beta chain, interleukin-12 subunit beta, natural killer cell stimulatory factor, 40 kD subunit, NKSF, NKSF2IL12, subunit p40 | |
| IL-12b | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. |
| ebdarokimab | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P29460 | |
| 3593 | |
| IgG1 | |
| Protein A purified | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title