missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL12B Antibody (I-1A4), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64693H
This item is not returnable.
View return policy
Description
IL12B Monoclonal specifically detects IL12B in Human, Porcine samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
| IL12B | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| IL12B | |
| Recombinant IL-12 | |
| 0.25 mL | |
| Cell Cycle and Replication, Cytokine Research, Immunology, Innate Immunity | |
| 3593 | |
| Human, Pig | |
| Purified |
| ELISA, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| I-1A4 | |
| ELISA, Immunohistochemistry, Immunohistochemistry-Frozen | |
| CLMF, CLMF p40, CLMF2, Cytotoxic lymphocyte maturation factor 40 kDa subunit, IL-12 subunit p40, IL-12BNK cell stimulatory factor chain 2, interleukin 12, p40, interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocytematuration factor 2, p40), interleukin-12 beta chain, interleukin-12 subunit beta, natural killer cell stimulatory factor, 40 kD subunit, NKSF, NKSF2IL12, subunit p40 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Recognizes the p40 subunit of human interleukin-12, and binds to both the free subunit and the p35/p40 heterodimer. The reacts with both stimulated and unstimulated monocytes in immunohistology. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction