missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL13RA1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17742414
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17742414 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17742414 Supplier Invitrogen™ Supplier No. PA5121016

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: HT-29, 293T. The target is usually found in the following locations: Membrane, Single-pass type I membrane protein. Immunogen sequence: TETQPPVTNL SVSVENLCTV IWTWNPPEGA SSNCSLWYFS HFGDKQDKKI APETRRSIEV PLNERICLQV GSQCSTNESE KPSILVEKCI SPPEGDPESA VTELQCIWHN LSYMKCSWLP GRNTSPDTNY TLYYWHRSLE KIHQCENIFR EGQYFGCSFD LTKVKDSSFE QHSVQIMVKD NAGKIKPSFN IVPLTSRVKP DPPHIKNLSF HNDDLYVQWE NPQNFISRCL FYEVEVNNSQ TETHNVFYVQ EAKCENPEFE RNVENTSCFM VPGVLPDTLN TVRIRVKTNK LCYEDDKLWS NWSQEMSIGK KRNST.

The protein encoded by this gene is a subunit of the interleukin 13 receptor. This subunit forms a receptor complex with IL4 receptor alpha, a subunit shared by IL13 and IL4 receptors. This subunit serves as a primary IL13-binding subunit of the IL13 receptor, and may also be a component of IL4 receptors. This protein has been shown to bind tyrosine kinase TYK2, and thus may mediate the signaling processes that lead to the activation of JAK1, STAT3 and STAT6 induced by IL13 and IL4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL13RA1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.34 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene IL13RA1
Gene Accession No. O09030, P78552
Gene Alias AI882074; Cancer/testis antigen 19; CD antigen CD213a1; CD213a1; CD213a1 antigen; CT19; IL13 receptor alpha-1 chain; IL-13 receptor subunit alpha-1; IL13R; IL-13R subunit alpha-1; IL-13r[a]; IL13RA; IL-13Ra; IL13RA1; IL-13RA1; IL-13R-alpha-1; interleukin 13 receptor subunit alpha 1; interleukin 13 receptor, alpha 1; interleukin 13 receptor, alpha 1-like; interleukin-13 receptor subunit alpha-1; Interleukin-13-binding protein; LOC100360218; Novel cytokine receptor 4; NR4
Gene Symbols IL13RA1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 29-343 of human IL13RA1 (NP_0015511).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16164, 3597
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.