Learn More
IL13RA2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11059-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is closely related to Il13RA1, a subuint of the interleukin 13 receptor complex. This protein binds IL13 with high affinity, but lacks cytoplasmic domain, and does not appear to function as a signal mediator. It is reported to play a role in the internalization of IL13.Specifications
| IL13RA2 | |
| Polyclonal | |
| Unconjugated | |
| IL13RA2 | |
| CD213A2, CT19, IL 13 receptor subunit alpha 2, IL 13R, IL 13R alpha 2, IL 13R subunit alpha 2, IL 13RA2, IL13BP, IL13R, IL13RA2, IL13Ralpha2, Interleukin 13 binding protein | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 16165, 171060, 3598 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O88786, Q14627, Q8VHK6 | |
| IL13RA2 | |
| IL13RA2 Fusion Protein Ag1534 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |