missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL17 (IL17A) Monoclonal Antibody (OTI3F6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies CF814521
This item is not returnable.
View return policy
Description
IL17 (IL17A) Monoclonal antibody specifically detects IL17 (IL17A) in Human samples. It is validated for ELISASpecifications
| IL17 (IL17A) | |
| Monoclonal | |
| See Label | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q16552 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 3605 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| ELISA | |
| OTI3F6 | |
| Unconjugated | |
| IL17A | |
| Ctla8; CTLA-8; cytokine; cytotoxic T-lymphocyte-associated antigen 8; cytotoxic T-lymphocyte-associated protein 8; il 17; Il17; IL-17; IL17A; IL-17a; ILN; Interleukin; interleukin 17; interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8); interleukin 17 precursor; interleukin 17A; interleukin-17; Interleukin17A; interleukin-17A; R-IL-17-A | |
| Full length human recombinant protein of human IL-17A (NP_002181) produced in 293 cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Lyophilized |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction