missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL17B Monoclonal Antibody (OTI3F7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811132S
This item is not returnable.
View return policy
Description
IL17B Monoclonal antibody specifically detects IL17B in Human samples. It is validated for Western BlotSpecifications
| IL17B | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9BS34 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 93474 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI3F7 | |
| Unconjugated | |
| ZNF670 | |
| 1110006O16Rik; 1700006N07Rik; Cytokine CX1; Cytokine Zcyto7; Cytokine-like protein ZCYTO7; gene for cytokine-like protein; IL17B; IL-17B; IL20; IL-20; interleukin 17B; interleukin 20; interleukin-17 beta; interleukin-17B; Interleukin20; Interleukin-20; neuronal interleukin-17 related factor; neuronal interleukin-17-related factor; NIRF; UNQ516/PRO1031; ZCYTO7 | |
| Full length human recombinant protein of human ZNF670 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction