missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL17RC Polyclonal Antibody
GREENER_CHOICE

Product Code. 15762041 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15762041 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15762041 Supplier Invitrogen™ Supplier No. PA554108

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: WDSDILCLPG DIVPAPGPVL APTHLQTELV LRCQKETDCD LCLRVAVHLA VHGHWEEPED EEKFGGAADS GVEEPRNASL QAQVVLSFQA YPTARCVLLE VQVPAALVQF GQSVGSVVYD CFEAALGSEV RIWSYTQPRY EKELNH Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 76%.

This gene encodes a single-pass type I membrane protein that shares similarity with the interleukin-17 receptor (IL-17RA). Unlike IL-17RA, which is predominantly expressed in hemopoietic cells, and binds with high affinity to only IL-17A, this protein is expressed in nonhemopoietic tissues, and binds both IL-17A and IL-17F with similar affinities. The proinflammatory cytokines, IL-17A and IL-17F, have been implicated in the progression of inflammatory and autoimmune diseases. Multiple alternatively spliced transcript variants encoding different isoforms have been detected for this gene, and it has been proposed that soluble, secreted proteins lacking transmembrane and intracellular domains may function as extracellular antagonists to cytokine signaling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL17RC
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene IL17RC
Gene Accession No. Q8NAC3
Gene Alias 1110025H02Rik; CANDF9; Gm19850; IL-17 receptor C; IL17F receptor; IL17RC; IL-17RC; IL17-RC; IL17Rhom; Il17rl; IL-17RL; IL17-RL; interleukin 17 receptor C; interleukin-17 receptor C; Interleukin-17 receptor homolog; interleukin-17 receptor-like protein; UNQ6118/PRO20040/PRO38901; ZcytoR14
Gene Symbols IL17RC
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human IL17RC. Recombinant protein control fragment (Product #RP-90444).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84818
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.