missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL18R1 Antibody (B-E43), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-14586H
This item is not returnable.
View return policy
Description
IL18R1 Monoclonal antibody specifically detects IL18R1 in Human samples. It is validated for ELISA
Specifications
| IL18R1 | |
| Monoclonal | |
| HRP | |
| CD218 antigen-like family member A, CD218a, CD218a antigen, CDw218a, cytokine receptor, IL-18R1, IL-18R-1, IL1R-rp, IL1RRPIL1 receptor-related protein, IL-1RrpIL18RA, interleukin 18 receptor 1, interleukin-18 receptor 1 | |
| Recombinant human IL18R1 | |
| 0.1 mL | |
| CD Markers | |
| 8809 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| ELISA | |
| B-E43 | |
| PBS | |
| Mouse | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction