missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL18R1 Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98223FR
This item is not returnable.
View return policy
Description
IL18R1 Polyclonal specifically detects IL18R1 in Mouse samples. It is validated for ELISA.
Specifications
| IL18R1 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 8809 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| DyLight 680 | |
| CD218 antigen-like family member A, CD218a, CD218a antigen, CDw218a, cytokine receptor, IL-18R1, IL-18R-1, IL1R-rp, IL1RRPIL1 receptor-related protein, IL-1RrpIL18RA, interleukin 18 receptor 1, interleukin-18 receptor 1 | |
| Produced in rabbits immunized with purified, recombinant IL18R1 (NP_032391.1; Met 1-Gly 326). | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction