Learn More
IL18R1 Antibody (MM0380-8H22), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-11679AF532
Product Code. 18163310
  • € 627.00 / 0.10mL
Description
IL18R1 Monoclonal specifically detects IL18R1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL18R1
Monoclonal
Alexa Fluor 532
50 mM sodium borate with 0.05% sodium azide
IL18R1
Human recombinant IL18R1 (extracellular domain)
0.1 mL
8809.0
Store at 4°C in the dark.
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
MM0380-8H22
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
CD218 antigen-like family member A, CD218a, CD218a antigen, CDw218a, cytokine receptor, IL-18R1, IL-18R-1, IL1R-rp, IL1RRPIL1 receptor-related protein, IL-1RrpIL18RA, interleukin 18 receptor 1, interleukin-18 receptor 1
Mouse
Protein G purified