missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL18R1 Recombinant Rabbit Monoclonal Antibody (BLR172J)
GREENER_CHOICE

Product Code. 17755899
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17755899 100 μL 100µL
17765899 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17755899 Supplier Invitrogen™ Supplier No. MA544529

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

IL18R1 Recombinant Monoclonal Antibody for Western Blot, Flow

IL18R1 is a cytokine receptor that belongs to the interleukin 1 receptor family. This receptor specifically binds interleukin 18 (IL18), and is essential for IL18 mediated signal transduction. IFN-alpha and IL12 are reported to induce the expression of this receptor in NK and T cells. The gene along with four other members of the interleukin 1 receptor family, including IL1R2, IL1R1, ILRL2 (IL-1Rrp2), and IL1RL1 (T1/ST2), form a gene cluster on chromosome 2q.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL18R1
Applications Flow Cytometry, Western Blot
Classification Recombinant Monoclonal
Clone BLR172J
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation BBS with 0.1% BSA and 0.09% sodium azide; pH 7.8-8.6
Gene Il18r1
Gene Accession No. Q13478
Gene Alias CD218 antigen-like family member A; CD218a; CDw218a; cytokine receptor; IL1 receptor-related protein; Il18r1; IL-18R1; IL-18R-1; IL18RA; Il18ralpha; IL-18R-alpha; IL18Ralpha2; IL1RRP; IL-1Rrp; IL1R-rp; interleukin 1 receptor related protein; interleukin 18 receptor 1; Interleukin-18; interleukin-18 receptor 1; Interleukin-18 receptor alpha
Gene Symbols Il18r1
Host Species Rabbit
Immunogen Recombinant protein representing the extracellular domain of Human Interleukin-18 receptor 1 (residues 25-329).
Purification Method Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8809
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.