Learn More
Invitrogen™ IL1F5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114747
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total IL1F5.
The protein encoded by this gene is a member of the interleukin 1 cytokine family. This cytokine was shown to specifically inhibit the activation of NF-kappaB induced by interleukin 1 family, member 6 (IL1F6). This gene and eight other interleukin 1 family genes form a cytokine gene cluster on chromosome 2. Two alternatively spliced transcript variants encoding the same protein have been reported.
Specifications
| IL1F5 | |
| Polyclonal | |
| Unconjugated | |
| IL36RN | |
| AI413231; FIL1; FIL1 delta; FIL1(DELTA); FIL1D; Fil1delta; IL-1 delta; IL-1 related protein 3; Il1f5; IL-1F5; IL1F5 (Canonical product IL-1F5a); IL-1F5 (IL-1HY1, FIL1-delta, IL-1RP3, IL-1L1, IL-1-delta); Il1h3; Il-1h3; IL1HY1; IL-1HY1; IL1L1; IL-1L1; IL-1ra homolog 1; IL-1-related protein 3; IL1RP3; IL-1RP3; IL36RA; IL-36Ra; IL36RN; Interleukin; interleukin 1 family, member 5; interleukin 1 family, member 5 (delta); interleukin 1 receptor antagonist homolog 1; interleukin 36 receptor antagonist; interleukin-1 delta; Interleukin-1 family member 5; interleukin-1 family member 5; interleukin-36 receptor antagonist protein; interleukin-1 homolog 3; Interleukin-1 HY1; interleukin-1 receptor antagonist homolog 1; Interleukin-1-like protein 1; interleukin-36 receptor antagonist protein; MGC29840; PSORP; PSORS14; UNQ1896/PRO4342 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 26525, 54450 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q9QYY1, Q9UBH0 | |
| Il1f5, IL36RN | |
| A synthesized peptide derived from human IL36RN(Accession Q9UBH0), corresponding to amino acid residues V83-L133. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |