missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL1F6 Monoclonal Antibody (OTI1D11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501428S
This item is not returnable.
View return policy
Description
IL1F6 Monoclonal antibody specifically detects IL1F6 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| IL1F6 | |
| Monoclonal | |
| 0.45 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9UHA7 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 27179 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Western Blot | |
| OTI1D11 | |
| Unconjugated | |
| IL36A | |
| FIL1; FIL1 epsilon; FIL1(EPSILON); Fil1e; Fil1epsilon; IL-1 epsilon; IL1(EPSILON); Il1e; Il1f6; IL-1F6; IL-1F6 (FIL-1-epsilon); Il1f9; Il1h1; IL-1H1; IL1RP2; Il36a; interleukin 1; interleukin 1 family, member 6; interleukin 1 family, member 6 (epsilon); interleukin 1 family, member 9; interleukin 1, epsilon; interleukin 36, alpha; interleukin-1 epsilon; Interleukin-1 family member 6; interleukin-1 homolog 1; Interleukin-36; Interleukin-36 alpha | |
| Full length human recombinant protein of human IL1F6 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction