Learn More
IL1F6 Mouse anti-Human, Clone: OTI1D11, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF501428
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The function of this protein has not been specifically defined.Specifications
| IL1F6 | |
| Monoclonal | |
| Unconjugated | |
| IL36A | |
| FIL1, FIL1(EPSILON), FIL1E, IL-1F6, IL1(EPSILON), IL1F6 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 27179 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI1D11 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q9UHA7 | |
| IL36A | |
| Full length human recombit protein of human IL1F6 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.