missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL1F8, Mouse anti-Human, Clone: 1E4, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00027177-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: MNPQREAAPKSYAIRDSRQMVWVLSGNSLIAAPLSRSIKPVTLHLIACRDTEFSDKEKGNMVYLGIKGKDLCLFCAEIQGKPTLQLKEKNIMDLYVEKKAQKPFLFFHNKEGSTSVFQSVSYPGWFIATSTTSGQPIFLTKERGITNNTNFYLDSVESpecifications
| IL1F8 | |
| Monoclonal | |
| Unconjugated | |
| interleukin 1 family, member 8 (eta) | |
| FIL1/FIL1-(ETA)/FIL1H/IL-1F8/IL-1H2/IL1-ETA/IL1H2/MGC126880/MGC126882 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 1E4 | |
| In 1x PBS, pH 7.4 | |
| NM_173178.1 | |
| IL1F8 | |
| IL1F8 (NP_775270.1, 1 a.a. ∼ 157 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 27177 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction