missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL1R1 Recombinant Rabbit Monoclonal Antibody (007), FITC
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA546873

Product Code. 17850556

  • € 431.00 / Pack of 100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Preparation: This antibody was obtained from a rabbit immunized with purified, recombinant Rat IL1R1 (Met1-Lys352) and conjugated with FITC under optimum conditions, the unreacted FITC was removed. Storage: This antibody can be stored at 2C-8C for twelve months without detectable loss of activity. Protected from prolonged exposure to light. Do not freeze! Sodium azide is toxic to cells and should be disposed of properly. Flush with large volumes of water during disposal.

The IL-1 system includes two agonists (IL-1 and IL-1), converting enzymes, antagonists, two receptors (IL-1 RI and IL-1 RII) and the IL-1 receptor accessory protein. Interleukin-1 signal is transduced through the type I receptor.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IL1R1
Recombinant Monoclonal
100 μg/mL
PBS with 0.5% BSA and 0.09% sodium azide
Q02955
Il1r1
Recombinant protein encoding Rat IL1R1 (Met1-Lys352).
100 Tests
Primary
Rat
Antibody
IgG
Flow Cytometry
007
FITC
Il1r1
antigen CD121a; CD121 alpha; CD121 antigen-like family member A; CD121a; CD121b; D2S1473; il1 receptor; il-1 receptor; IL-1 receptor alpha chain; IL1R; il-1r; IL1R1; IL-1R1; IL-1R-1; Il1r-1; IL1RA; IL-1R-alpha; IL1RT1; IL-1RT1; IL-1RT-1; IL-iR; interleukin 1 beta receptor type 1 short transmembrane form; interleukin 1 beta receptor type 1 soluble form; interleukin 1 bete receptor type 1 long transmembrane form; interleukin 1 receptor alpha, type I; interleukin 1 receptor type 1; interleukin 1 receptor, type I; interleukin-1 receptor; interleukin-1 receptor alpha; Interleukin-1 receptor type 1; Interleukin-1 receptor type 1, membrane form; Interleukin-1 receptor type 1, soluble form; Interleukin-1 receptor type I; mIL-1R1; p80; sIL-1R1
Rabbit
Protein A
RUO
25663
4°C, store in dark, DO NOT FREEZE!
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.