missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL1RAPL1 Antibody (B-L43), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-18110DL594
This item is not returnable.
View return policy
Description
IL1RAPL1 Monoclonal antibody specifically detects IL1RAPL1 in Human samples. It is validated for Flow Cytometry, ELISA
Specifications
| IL1RAPL1 | |
| Monoclonal | |
| DyLight 594 | |
| IL1R8IL1RAPL, IL-1-RAPL-1, IL-1RAPL-1, IL1RAPL-1, interleukin 1 receptor accessory protein-like 1, interleukin 1 receptor-8, interleukin-1 receptor accessory protein-like 1, mental retardation, X-linked 10, MRX10, MRX21, MRX34, Oligophrenin-4, OPHN4mental retardation, X-linked 21, Three immunoglobulin domain-containing IL-1 receptor-related 2, TIGIRR-2mental retardation, X-linked 34, X-linked interleukin-1 receptor accessory protein-like 1 | |
| Recombinant human IL1RAPL1. Hybridoma: Myeloma X63/AG.8653 x Balb/c spleen cells | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA | |
| B-L43 | |
| 50mM Sodium Borate | |
| Mouse | |
| Affinity purified | |
| RUO | |
| 11141 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction