missing translation for 'onlineSavingsMsg'
Learn More

IL1RAPL2 Antibody, Novus Biologicals™

Product Code. p-200064981 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18613556 100 μg 100µL
18648045 50 μg 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18613556 Supplier Novus Biologicals Supplier No. NBP298932100ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IL1RAPL2 Polyclonal antibody specifically detects IL1RAPL2 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL1RAPL2
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution ELISA 1:5000-1:10000
Formulation 0.2 um filtered solution in PBS
Gene Alias IL-1R-9, IL1R9IL-1 receptor accessory protein-like 2, IL-1R9IL-1RAPL-2, IL-1-RAPL-2, IL1RAPL-2IL1RAPL-2-related protein, interleukin 1 receptor 9, interleukin 1 receptor accessory protein-like 2, Three immunoglobulin domain-containing IL-1 receptor-related 1, TIGIRR-1Interleukin-1 receptor 9, X-linked interleukin-1 receptor accessory protein-like 2
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human IL1RAPL2 (Accession#: NP_059112.1; Met 1-Glu 356)
Purification Method Antigen and protein A Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26280
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.