missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL1RAPL2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104273
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total IL-1R9.
The protein encoded by this gene is a member of the interleukin 1 receptor family. This protein is similar to the interleukin 1 accessory proteins, and is most closely related to interleukin 1 receptor accessory protein-like 1 (IL1RAPL1). This gene and IL1RAPL1 are located at a region on chromosome X that is associated with X-linked non-syndromic mental retardation. [provided by RefSeq, Jul 2008].
Specifications
| IL1RAPL2 | |
| Polyclonal | |
| Unconjugated | |
| IL1RAPL2 | |
| IL-1 receptor accessory protein-like 2; IL1R9; IL-1R9; IL-1R-9; IL1RAPL2; IL-1-RAPL-2; IL-1RAPL-2; IL1RAPL-2; IL1RAPL-2-related protein; interleukin 1 receptor 9; interleukin 1 receptor accessory protein like 2; interleukin 1 receptor accessory protein-like 2; Interleukin-1 receptor 9; RGD1561761; three immunoglobulin domain-containing IL-1 receptor-related 1; TIGIRR-1; X-linked interleukin-1 receptor accessory protein-like 2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 26280, 300913, 60367 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9ERS6, Q9NP60 | |
| IL1RAPL2 | |
| A synthesized peptide derived from human IL1RAPL2(Accession Q9NP60), corresponding to amino acid residues S144-Q194. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction