Learn More
IL1RN Monoclonal Antibody (OTI2D9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA803293
Encompass_Preferred
 View more versions of this product
Product Code. 17502273
  • € 540.87 / 100µL
Description
IL1RA (IL1RN) protein is a cytokine receptor that belongs to the interleukin 1 receptor family. IL1RA is a receptor for interleukin alpha (IL1A), interleukin beta (IL1B), and interleukin 1 receptor, type I(IL1R1/IL1RA). It is an important mediator involved in many cytokine induced immune and inflammatory responses. This gene along with interleukin 1 receptor, type II (IL1R2), interleukin 1 receptor-like 2 (IL1RL2), and interleukin 1 receptor-like 1 (IL1RL1) form a cytokine receptor gene cluster in a region mapped to chromosome 2q12.
Specifications
Specifications
IL1RN
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P18510
IL1RN
Full length human recombit protein of human IL1RN produced in E.coli.
100 μL
Primary
Human
Antibody
IgG2b
Western Blot
OTI2D9
Unconjugated
IL1RN
DIRA, ICIL-1RA, IL-1RN, IL-1ra, IL-1ra3, IL1F3, IL1RA, IRAP, MVCD4
Mouse
Affinity Chromatography
RUO
3557
-20° C, Avoid Freeze/Thaw Cycles