Learn More
IL1RN Mouse anti-Human, Clone: OTI2D2, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF803483
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
IL1RA (IL1RN) protein is a cytokine receptor that belongs to the interleukin 1 receptor family. IL1RA is a receptor for interleukin alpha (IL1A), interleukin beta (IL1B), and interleukin 1 receptor, type I(IL1R1/IL1RA). It is an important mediator involved in many cytokine induced immune and inflammatory responses. This gene along with interleukin 1 receptor, type II (IL1R2), interleukin 1 receptor-like 2 (IL1RL2), and interleukin 1 receptor-like 1 (IL1RL1) form a cytokine receptor gene cluster in a region mapped to chromosome 2q12.Specifications
| IL1RN | |
| Monoclonal | |
| Unconjugated | |
| IL1RN | |
| DIRA, ICIL-1RA, IL-1RN, IL-1ra, IL-1ra3, IL1F3, IL1RA, IRAP, MVCD4 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 3557 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI2D2 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P18510 | |
| IL1RN | |
| Full length human recombit protein of human IL1RN produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.