missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL2 Monoclonal Antibody (OTI5A9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810802S
This item is not returnable.
View return policy
Description
IL2 Monoclonal antibody specifically detects IL2 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| IL2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q15109 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 177 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Immunohistochemistry (Paraffin) | |
| OTI5A9 | |
| Unconjugated | |
| AGER | |
| aldesleukin; cytokine; H-IL-2; il 2; Il2; Il-2; ILN; Interleukin; interleukin 2; interleukin 2 precursor; interleukin 2 variant IL2delta2; Interleukin2; Interleukin-2; interleukin-2 precursor; interleukin-2 precursor protein; interleukin-2; interleukin 2; Involved in regulation of T cell clonal expansion; involved in regulation of T-cell clonal expansion; lymphokine; M-IL-2; patial sequence of Interleukin-2; POIL2; porcine Interleukin 2; T cell growth factor; T-cell growth factor; T-cell growth factor (TCGF); TCGF | |
| Human recombinant protein fragment corresponding to amino acids 205-419 of human RAGE produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction