missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL2 Monoclonal Antibody (OTI5A9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810802
This item is not returnable.
View return policy
Description
Interleukin 2 (IL-2) is an immuno-modulatory cytokine that is important for the proliferation of activated T cells, differentiation of B cells, natural killer cells, monocytes and macrophages. IL-2 signals through the IL-2 receptor (IL-2R), a heterotrimeric protein complex whose gamma chain is also shared by interleukin 4 (IL-4) and interleukin 7 (IL-7). The expression of the IL-2 gene in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. The targeted disruption of a gene similar to IL-2 in mice leads to an ulcerative colitis-like disease that suggests an essential role of this gene in the immune response to antigenic stimuli.Specifications
| IL2 | |
| Monoclonal | |
| Unconjugated | |
| IL2 | |
| IL-2, TCGF, lymphokine | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 3558 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| OTI5A9 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P60568 | |
| IL2 | |
| Human recombit protein fragment corresponding to amino acids 21-153 of human IL2 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction