Learn More
IL20 Mouse anti-Human, Clone: OTI5E10, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF803409
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Cytokines that signal through Class II receptors form a distinct family that includes the interferons and interleukin 10 (IL-10). Recent identification of several IL-10 homologs has defined a cytokine subfamily that includes AK155, IL-19, IL-20, IL-22, and IL-24. IL-20 belongs to the IL-10 family and plays a role in skin inflammation and the development of hematopoietic cells. Little is known about its other biological functions and clinical implications. These cytokines display pro-inflammatory activities in different tissues, including skin.Specifications
| IL20 | |
| Monoclonal | |
| Unconjugated | |
| IL20 | |
| IL-20, IL10D, ZCYTO10 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 50604 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI5E10 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q9NYY1 | |
| IL20 | |
| Human recombit protein fragment corresponding to amino acids 25-176 of human IL20 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.