missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL20RA Monoclonal Antibody (OTI6F8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811647
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a receptor for interleukin 20 (IL20), a cytokine that may be involved in epidermal function. The receptor of IL20 is a heterodimeric receptor complex consisting of this protein and interleukin 20 receptor beta (IL20B). This gene and IL20B are highly expressed in skin. The expression of both genes is found to be upregulated in Psoriasis.Specifications
| IL20RA | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9UHF4 | |
| IL20RA | |
| Human recombit protein fragment corresponding to amino acids 272-553 of human IL20RA produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI6F8 | |
| Unconjugated | |
| IL20RA | |
| CRF2-8, IL-20R-alpha, IL-20R1, IL-20RA | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 53832 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction