missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL20RA Monoclonal Antibody (OTI7H7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811623S
This item is not returnable.
View return policy
Description
IL20RA Monoclonal antibody specifically detects IL20RA in Human samples. It is validated for Western BlotSpecifications
| IL20RA | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O15479 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 4113 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI7H7 | |
| Unconjugated | |
| MAGEB2 | |
| class II cytokine receptor ZCYTOR7; CRF2-8; Cytokine receptor class-II member 8; Cytokine receptor family 2 member 8; E230031K19; IL-20 receptor subunit alpha; IL-20R1; IL20RA; IL-20RA; IL-20R-alpha; interleukin 20 receptor alpha subunit; interleukin 20 receptor subunit alpha; interleukin 20 receptor, alpha; interleukin-20 receptor I; interleukin-20 receptor subunit alpha; PRO1315; UNQ681; UNQ681/PRO1315; ZcytoR7 | |
| Human recombinant protein fragment corresponding to amino acids 1-224 of human MAGEB2 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction