Learn More
IL20RA Mouse anti-Human, Clone: OTI6F8, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand:  Origene Technologies CF811647
Encompass_Preferred
Product Code. 17631902
  • € 726.00 / 100µg
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene is a receptor for interleukin 20 (IL20), a cytokine that may be involved in epidermal function. The receptor of IL20 is a heterodimeric receptor complex consisting of this protein and interleukin 20 receptor beta (IL20B). This gene and IL20B are highly expressed in skin. The expression of both genes is found to be upregulated in Psoriasis.
Specifications
Specifications
IL20RA
Monoclonal
Unconjugated
IL20RA
CRF2-8, IL-20R-alpha, IL-20R1, IL-20RA
Mouse
Affinity Chromatography
RUO
53832
-20° C, Avoid Freeze/Thaw Cycles
Lyophilized
Western Blot
OTI6F8
PBS with 8% trehalose and no preservative; pH 7.3
Q9UHF4
IL20RA
Human recombit protein fragment corresponding to amino acids 272-553 of human IL20RA produced in E.coli.