missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL20RB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584623
This item is not returnable.
View return policy
Description
Immunogen sequence: QFEFLVAYWR REPGAEEHVK MVRSGGIPVH LETMEPGAAY CVKAQTFVKA IGRYSAFSQT ECVEVQGE Highest antigen sequence indentity to the following orthologs: Mouse - 85%, Rat - 76%.
IL20RB and IL20RA (MIM 605620) form a heterodimeric receptor for interleukin-20 (IL20; MIM 605619) (Blumberg et al., 2001 [PubMed 11163236]).[supplied by OMIM, Feb 2009].
Specifications
| IL20RB | |
| Polyclonal | |
| Unconjugated | |
| Il20rb | |
| AV228068; DIRS1; Fibronectin type III domain containing 6; FNDC6; Gm186; IL-20 receptor subunit beta; IL20R2; IL-20R2; IL20RB; IL-20RB; IL-20R-beta; interleukin 20 receptor beta; interleukin 20 receptor beta subunit; interleukin 20 receptor subunit beta; interleukin-20 receptor II; interleukin-20 receptor subunit beta; RGD1566151; UNQ557/PRO1114 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 53833 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6UXL0 | |
| Il20rb | |
| Recombinant protein corresponding to Human IL20RB. Recombinant protein control fragment (Product #RP-108495). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction