Learn More
Invitrogen™ IL21R Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596690
Description
Immunogen sequence: GVSMPRGWAA PLLLLLLQGG WGCPDLVCYT DYLQTVICIL EMWNLHPSTL TLTWQDQYEE LKDEATSCSL HRSAHNATHA TYTCHMDVFH FMADDIFSVN ITDQSGNYSQ ECGSFLLAES IKPAPPFNVT VTFSGQYNIS WRSDYEDPAF YMLKGKLQYE LQYRNRGDPW AVSPRRKLIS VDSRSVSLLP LEFRKDSSYE LQVRAGPMPG SSYQGTWSEW S; Positive Samples: U-87MG, Mouse spleen, Mouse thymus; Cellular Location: Membrane, Single-pass type I membrane protein.
The protein encoded by this gene is a cytokine receptor for interleukin 21 (IL21). It belongs to the type I cytokine receptors, and has been shown to form a heterodimeric receptor complex with the common gamma-chain, a receptor subunit also shared by the receptors for interleukin 2 (IL2) and interleukin 5 (IL5). This receptor transduces the growth promoting signal of IL21, and is important for the proliferation and differentiation of T cells, B cells, and natural killer (NK) cells. The ligand binding of this receptor leads to the activation of multiple downstream signaling molecules, including JAK1, JAK3, STAT1, and STAT3. Knockout studies of a similar gene in mouse suggest a role for this gene in regulating immunoglobulin production. Three alternatively spliced transcript variants encoding the same protein have been described.
Specifications
| IL21R | |
| Polyclonal | |
| Unconjugated | |
| Il21r | |
| CD360; IL-21 receptor; Il21r; IL-21R; interleukin 21 receptor; interleukin-21 receptor; LR-beta; lymphocyte receptor beta; MGC10967; NILR; novel cytokine receptor 8; Novel interleukin receptor; NR8; UNQ3121/PRO10273 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 50615, 60504 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9HBE5, Q9JHX3 | |
| Il21r | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 20-240 of human IL21R (NP_8515654). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.