missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL22RA2 Polyclonal Antibody
Description
Recommended positive controls: Human spleen or tonsil..
IL22, a homolog of IL10, is secreted by T cells and binds to and signals through the class II cytokine receptor family (CRF) heterodimer IL22RA /IL10RB. IL22 induces the production of acute-phase reactants.
Specifications
Specifications
| Antigen | IL22RA2 |
| Applications | ELISA, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% BSA and 0.1% sodium azide |
| Gene | IL22RA2 |
| Gene Accession No. | Q7TNI4, Q80XF5, Q969J5 |
| Gene Alias | CRF2-10; CRF2-S1; CRF2X; Cytokine receptor class-II member 10; Cytokine receptor family 2 member 10; cytokine receptor family II soluble 1; cytokine receptor family type 2, soluble 1; IL-22 receptor subunit alpha-2; IL22BP; IL-22BP; Il22ra2; IL-22RA2; IL-22R-alpha-2; interleukin 22 receptor subunit alpha 2; interleukin 22 receptor, alpha 2; interleukin 22-binding protein; interleukin-22 receptor subunit alpha-2; Interleukin-22-binding protein; UNQ5793/PRO19598/PRO19822; ZcytoR16 |
| Gene Symbols | IL22RA2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?