missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL24 Monoclonal Antibody (OTI4H4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809452S
This item is not returnable.
View return policy
Description
IL24 Monoclonal antibody specifically detects IL24 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| IL24 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9H6K4 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 80207 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4H4 | |
| Unconjugated | |
| OPA3 | |
| C49A; Cytokine-like protein Mob-5; FISP; IL 24; IL10B; Il24; IL-24; IL-4-induced secreted protein; interleukin 24; interleukin-24; Mda7; MDA-7; melanocyte-associated Mda-7; melanoma differentiation-associated gene 7 protein; Mob5; ST16; suppression of tumorigenicity 16 (melanoma differentiation); Suppression of tumorigenicity 16 protein; Th2-specific cytokine FISP | |
| Full length human recombinant protein of human OPA3 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction