Learn More
IL24 Mouse anti-Human, Clone: OTI3C5, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF809390
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Interleukin 24 (IL-24)/melanoma differentiation-associated gene-7 (mda-7) encodes a secreted protein that exhibits significant homology to the interleukin 10 (IL-10) family of cytokines. The human IL-24 is secreted by activated peripheral blood mononuclear cells and binds two heterodimeric receptors, IL-22R1/IL-20R2 and IL-20R1/IL-20R2 resulting in the activation of JAK/STAT signaling pathways. Mda-7/IL-24, selectively induces growth suppression, apoptosis and radiosensitization in maligt gliomas in a p53-independent manner.Specifications
| IL24 | |
| Monoclonal | |
| Unconjugated | |
| IL24 | |
| C49A, FISP, IL10B, MDA7, MOB5, ST16 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 11009 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3C5 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q13007 | |
| IL24 | |
| Human recombit protein fragment corresponding to amino acids 52-206 of human IL24 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.