missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Il2ra, Rat anti-Mouse, Clone: PC61, Abnova™
Rat Monoclonal Antibody
Brand: Abnova
This item is not returnable.
View return policy
Specifications
| Il2ra | |
| Monoclonal | |
| Unconjugated | |
| In PBS, pH 7.4 (protein stabilizer, 0.09% sodium azide). | |
| CD25/Il2r/Ly-43/MGC130561 | |
| Rat | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG1 |
| Flow Cytometry | |
| PC61 | |
| Flow Cytometry (≤ 1 ug/106 cells) The optimal working dilution should be determined by the end user. | |
| interleukin 2 receptor, alpha chain | |
| Il2ra | |
| IL2-dependent cytolytic mouse T-cell clone B6.1. | |
| RUO | |
| 16184 | |
| Store in the dark at 4°C. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction