missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL2RB Mouse anti-Human, Clone:1G11H3, Abnova™
Mouse monoclonal antibody raised against partial recombinant human IL2RB.
Brand: Abnova MAB17990.100ug
This item is not returnable.
View return policy
Description
The interleukin 2 receptor, which is involved in T cell-mediated immune responses, is present in 3 forms with respect to ability to bind interleukin 2. The low affinity form is a monomer of the alpha subunit and is not involved in signal transduction. The intermediate affinity form consists of an alpha/beta subunit heterodimer, while the high affinity form consists of an alpha/beta/gamma subunit heterotrimer. Both the intermediate and high affinity forms of the receptor are involved in receptor-mediated endocytosis and transduction of mitogenic signals from interleukin 2. The protein encoded by this gene represents the beta subunit and is a type I membrane protein. [provided by RefSeq]Specifications
| IL2RB | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Flow Cytometry (1:200-1:400) Western Blot (1:100-1:500) The optimal working dilution should be determined by the end user. |
|
| IL2RB | |
| Mouse | |
| 100 μg | |
| Human | |
| Antibody | |
| IgG2b |
| ELISA, Flow Cytometry, Western Blot | |
| 1G11H3 | |
| interleukin 2 receptor, beta | |
| In PBS (0.05% sodium azide) | |
| CD122, P70-75 | |
| Recombinant protein corresponding to amino acids 27-240 of human IL2RB. | |
| 3560 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction