Learn More
IL34 Mouse anti-Human, Clone: OTI4A6, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF811727
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
IL-34 is secreted as a dimeric protein and currently belongs to no known cytokine family. It signals via the M-CSF Receptor, also known as CSF-1R and CD115, which is shared with M-CSF. IL-34 has no amino acid sequence homology to M-CSF, but they exhibit similarities in secondary structure and share overlapping functions. IL-34 has been shown to promote myeloid survival, proliferation, and differentiation, and to play a role in osteoclast formation. IL-34 expression has been detected in various tissues and is most abundant in the spleen.Specifications
| IL34 | |
| Monoclonal | |
| Unconjugated | |
| IL34 | |
| C16orf77, IL-34 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 146433 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI4A6 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q6ZMJ4 | |
| IL34 | |
| Human recombit protein fragment corresponding to amino acids 77-242 of human IL34 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.