missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL34 Mouse anti-Human, Clone: OTI5E1, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811737S
This item is not returnable.
View return policy
Description
IL34 Monoclonal antibody specifically detects IL34 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| IL34 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q6ZMJ4 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 146433 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI5E1 | |
| Unconjugated | |
| Il34 | |
| 2010004A03Rik; AI593503; C16orf77; IL34; IL-34; ILN; Interleukin; interleukin 34; Interleukin34; interleukin-34 | |
| Human recombinant protein fragment corresponding to amino acids 77-242 of human IL34 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction