missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL6 Monoclonal Antibody (OTI2B3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies CF813867
This item is not returnable.
View return policy
Description
IL6 Monoclonal antibody specifically detects IL6 in Human samples. It is validated for ELISASpecifications
| IL6 | |
| Monoclonal | |
| See Label | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P02671 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 2243 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| ELISA | |
| OTI2B3 | |
| Unconjugated | |
| FGA | |
| B-cell differentiation factor; B-cell hybridoma growth factor; B-cell stimulatory factor 2; BSF2; BSF-2; CDF; CHIL-6; CTL differentiation factor; cytokine; HGF; H-IL-6; HSF; hybridoma growth factor; Ifnb2; IFN-beta-2; Il6; IL-6; ILg6; ILN; interferon beta-2; Interleukin; interleukin 6; Interleukin 6 (interferon, beta 2); interleukin 6 precursor; interleukin BSF-2; interleukin HP-1; Interleukin6; Interleukin-6; interleukin-6 precursor; interleukin-6 protein; M-IL-6; prointerleukin 6; R-IL-6 | |
| Native Human D-Dimer protein | |
| 100 μg | |
| Primary | |
| Human | |
| Lyophilized |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction