missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL6R Antibody (B-F19) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-18095JF525
This item is not returnable.
View return policy
Description
IL6R Monoclonal antibody specifically detects IL6R in Human samples. It is validated for Flow Cytometry,ELISA,Immunohistochemistry
Specifications
| IL6R | |
| Monoclonal | |
| Janelia Fluor 525 | |
| CD126, CD126 antigen, gp80, IL-6 receptor subunit alpha, IL-6R 1, IL-6R subunit alpha, IL-6R-1, IL6RA, IL-6RA, IL-6R-alpha, interleukin 6 receptor, interleukin 6 receptor alpha subunit, interleukin-6 receptor subunit alpha, Membrane glycoprotein 80, MGC104991 | |
| gp80 transfected CHO cell line. Hybridoma: Myeloma X63/AG.8653 x Balb/c spleen cells | |
| 0.1 mL | |
| Cancer, Cytokine Research, Immunology, Innate Immunity | |
| 3570 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry, ELISA, Immunohistochemistry | |
| B-F19 | |
| 50mM Sodium Borate | |
| Mouse | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction