missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL6R Antibody (B-R6), Allophycocyanin/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64770APCCY7
This item is not returnable.
View return policy
Description
IL6R Monoclonal specifically detects IL6R in Human samples. It is validated for Flow Cytometry.
Specifications
| IL6R | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| IL6R | |
| CHO cells transfected with cDNA of the IL-6 receptor | |
| 0.1 mL | |
| Primary | |
| Recognizes soluble and membranous human IL-6R. This has been shown to inhibit IL-6 mediated proliferation of XG-1 cells, and partially blocks binding of IL-6 to its receptor. Removal of Sodium azide is recommended prior to use in functional assays. | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| B-R6 | |
| Flow Cytometry | |
| CD126, CD126 antigen, gp80, IL-6 receptor subunit alpha, IL-6R 1, IL-6R subunit alpha, IL-6R-1, IL6RA, IL-6RA, IL-6R-alpha, interleukin 6 receptor, interleukin 6 receptor alpha subunit, interleukin-6 receptor subunit alpha, Membrane glycoprotein 80, MGC104991 | |
| Mouse | |
| Protein A or G purified | |
| Cancer, Cytokine Research, Immunology, Innate Immunity | |
| 3570.0 | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction