missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL6R Antibody (D7715A7), HRP, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP3-12010H
This item is not returnable.
View return policy
Description
IL6R Monoclonal antibody specifically detects IL6R in Mouse samples. It is validated for Flow Cytometry, Neutralization
Specifications
| IL6R | |
| Monoclonal | |
| HRP | |
| PBS | |
| Rat | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry, Neutralization | |
| D7715A7 | |
| Flow Cytometry, Block/Neutralize | |
| CD126, CD126 antigen, gp80, IL-6 receptor subunit alpha, IL-6R 1, IL-6R subunit alpha, IL-6R-1, IL6RA, IL-6RA, IL-6R-alpha, interleukin 6 receptor, interleukin 6 receptor alpha subunit, interleukin-6 receptor subunit alpha, Membrane glycoprotein 80, MGC104991 | |
| This antibody was raised by immunising Lewis rats with OKT4 cells. | |
| 0.1 mL | |
| Cancer, Cytokine Research, Immunology, Innate Immunity | |
| 3570 | |
| Store at 4C in the dark. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction