missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL6R Antibody (OTI2F4), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71029B
This item is not returnable.
View return policy
Description
IL6R Monoclonal specifically detects IL6R in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| IL6R | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| IL6R | |
| Full length recombinant protein of human IL6R (NP_000556) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI2F4 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| CD126, CD126 antigen, gp80, IL-6 receptor subunit alpha, IL-6R 1, IL-6R subunit alpha, IL-6R-1, IL6RA, IL-6RA, IL-6R-alpha, interleukin 6 receptor, interleukin 6 receptor alpha subunit, interleukin-6 receptor subunit alpha, Membrane glycoprotein 80, MGC104991 | |
| Mouse | |
| Affinity Purified | |
| Cancer, Cytokine Research, Immunology, Innate Immunity | |
| 3570.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction