Learn More
IL6R Mouse anti-Human, Clone: OTI2F4, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF506859
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
IL6R (interleukin-6 receptor subunit alpha) is a part of the receptor for interleukin 6. The IL-6 receptor complex is composed of two membrane glycoproteins: the low affinity receptor and the signal transducing component. Dysregulated production of IL6 and this receptor are implicated in the pathogenesis of many diseases, such as multiple myeloma, autoimmune diseases and prostate cancer.Specifications
| IL6R | |
| Monoclonal | |
| Unconjugated | |
| IL6R | |
| CD126, IL-6R-1, IL-6RA, IL6Q, IL6RA, IL6RQ, gp80 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 3570 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI2F4 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P08887 | |
| IL6R | |
| Full length human recombit protein of human IL6R produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.