missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ILF2 Antibody (OTI6F1), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71036G
This item is not returnable.
View return policy
Description
ILF2 Monoclonal specifically detects ILF2 in Human, Mouse, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| ILF2 | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ILF2 | |
| Full-length protein expressed in 293T cell transfected with human ILF2 expression vector. | |
| 0.1 mL | |
| 3608.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI6F1 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| interleukin enhancer binding factor 2, 45kD, interleukin enhancer binding factor 2, 45kDa, interleukin enhancer-binding factor 2, MGC8391, NF45PRO3063, Nuclear factor of activated T-cells 45 kDa, nuclear factor of activated T-cells, 45-kDa | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Canine | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction