missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ILF3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
€ 386.00 - € 529.00
Specifications
| Antigen | ILF3 |
|---|---|
| Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry 1:2500 - 1:5000, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:2500 - 1:5000 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18476220
|
Novus Biologicals
NBP1-84056-25ul |
25 μL |
€ 386.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18265936
|
Novus Biologicals
NBP1-84056 |
0.1 mL |
€ 529.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ILF3 Polyclonal specifically detects ILF3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| ILF3 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 3609 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:VMAKHSSVYPTQEELEAVQNMVSHTERALKAVSDWIDEQEKGSSEQAESDNMDVPPEDDSKEGAGEQKTEHMTRTLRGVMRVGLVAKGLLLKGDLDLELVLLCKEKPTTALLDKVADNLAIQLAAVTEDKYEILQSVDDAAIVIK | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 0.4 ug/ml, Immunohistochemistry 1:2500 - 1:5000, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:2500 - 1:5000 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Double-stranded RNA-binding protein 76, double-stranded RNA-binding protein, 76 kD, DRBF, DRBP76M-phase phosphoprotein 4, dsRNA binding protein NFAR-2/MPP4, interleukin enhancer binding factor 3, 90kDa, interleukin enhancer-binding factor 3, MPHOSPH4interleukin enhancer binding factor 3, 90kD, MPP4MMP4, NF90CBTF, NFAR-1, NFAR2, NFARNF110, NF-AT-90, Nuclear factor associated with dsRNA, Nuclear factor of activated T-cells 90 kDa, nuclear factor of activated T-cells, 90 kD, TCP110, TCP80, Translational control protein 80 | |
| ILF3 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title