missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ILF3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA541536
This item is not returnable.
View return policy
Description
Peptide sequence: IFVNDDRHVM AKHSSVYPTQ EELEAVQNMV SHTERALKAV SDWIDEQEKG Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.
UBA6 (UBE1L2, MOP-4)is a novel E1 enzyme specific for ubiquitin and belongs to the ubiquitin-activating E1 family. It is a part of the cascade to attach ubiquitin (Ub) covalently to substrate proteins. UBA6 activates ubiquitin by first adenylating its carboxy-terminal glycine residue with ATP and then linking this residue to the side chain of a cysteine residue in E1, yielding an ubiquitin-E1 thioester and free AMP. Its isoforms influence spermatogenesis and male fertility. UBA6 is highly expressed in human monocytes.
Specifications
| ILF3 | |
| Polyclonal | |
| Unconjugated | |
| ILF3 | |
| CBTF; Double-stranded RNA-binding protein 76; double-stranded RNA-binding protein, 76 kD; DRBF; DRBP76; dsRNA binding protein NFAR-2/MPP4; Ilf3; ilf3 protein; ilf3b; interleukin enhancer binding factor 3; interleukin enhancer binding factor 3, 90kD; interleukin enhancer binding factor 3, 90kDa; interleukin enhancer binding factor 3b; interleukin enhancer-binding factor 3; Interleukin enhancer-binding factor 3 homolog; M phase phosphoprotein 4; MBII-26; MMP4; M-phase phosphoprotein 4; MPHOSPH4; MPP4; NF110; NF110b; NF90; NF90a; NF90b; NFAR; NFAR-1; NFAR2; NF-AT-90; nuclear factor 90; nuclear factor associated with dsRNA; Nuclear factor of activated T-cells 90 kDa; nuclear factor of activated T-cells, 90 kD; TCP110; TCP80; Translational control protein 80; wu:fb37d07; wu:fb94b02; zgc:77030 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 3609, 84472 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q12906, Q9JIL3 | |
| ILF3 | |
| Synthetic peptide directed towards the N-terminal of human ILF3 (aa 6-55). | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction