Learn More
Invitrogen™ ILF3 Recombinant Rabbit Monoclonal Antibody (7H8)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA549922
Product Code. 17999452
  • € 446.00 / 100µL
Description
ILF3 Recombinant Monoclonal Antibody for ICC/IF, IHC (P), ELISA
UBA6 (UBE1L2, MOP-4)is a novel E1 enzyme specific for ubiquitin and belongs to the ubiquitin-activating E1 family. It is a part of the cascade to attach ubiquitin (Ub) covalently to substrate proteins. UBA6 activates ubiquitin by first adenylating its carboxy-terminal glycine residue with ATP and then linking this residue to the side chain of a cysteine residue in E1, yielding an ubiquitin-E1 thioester and free AMP. Its isoforms influence spermatogenesis and male fertility. UBA6 is highly expressed in human monocytes.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ILF3
Recombinant Monoclonal
0.85 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q12906
ILF3
A synthesized peptide derived from human ILF3.
100 μL
Primary
Human
Antibody
IgG
ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry
7H8
Unconjugated
ILF3
CBTF; Double-stranded RNA-binding protein 76; double-stranded RNA-binding protein, 76 kD; DRBF; DRBP76; dsRNA binding protein NFAR-2/MPP4; Ilf3; ilf3 protein; ilf3b; interleukin enhancer binding factor 3; interleukin enhancer binding factor 3, 90kD; interleukin enhancer binding factor 3, 90kDa; interleukin enhancer binding factor 3b; interleukin enhancer-binding factor 3; Interleukin enhancer-binding factor 3 homolog; M phase phosphoprotein 4; MBII-26; MMP4; M-phase phosphoprotein 4; MPHOSPH4; MPP4; NF110; NF110b; NF90; NF90a; NF90b; NFAR; NFAR-1; NFAR2; NF-AT-90; nuclear factor 90; nuclear factor associated with dsRNA; Nuclear factor of activated T-cells 90 kDa; nuclear factor of activated T-cells, 90 kD; TCP110; TCP80; Translational control protein 80; wu:fb37d07; wu:fb94b02; zgc:77030
Rabbit
Affinity chromatography
RUO
3609
-20°C or -80°C if preferred