Learn More
Cytiva Hot Start Master Mix
Product Code. 10269384 Shop All Cytiva Products
Click to view available options
:
Hot Start Master Mix
Product Code. 10269384
Brand: Cytiva 25150001
A ready-to-use 2X mastermix providing robust and reliable performance for demanding PCR applications in which high specificity and high sensitivity are essential to success
  • Convenient, ready-to-use mix saves time and reduces experimental variability
  • For any standard PCR reaction, such as multiplex PCR, and reactions exhibiting nonspecific amplification or primer-dimer formation
  • Primer sequestration effectively reduces any nonspecific priming or primer-dimer formation
  • Recombinant activator protein avoids the risk of contamination from a mammalian source associated with hot start antibodies
  • Avoids the risk of contamination from a mammalian source associated with hot start antibodies
  • Quick hot start activation preserves sample integrity by avoiding extensive precycling heat treatment
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 2X Concentrate solution of 20mM Tris HCl (pH 8.6), 100mM KCl, 3mM MgCl2, 0.4mM dNTPs (dATP, dCTP, dGTP, dTTP), 50U/mL Taq DNA Polymerase, hot start activator protein, stabilizers. Store at -20°C
Description Hot Start Master Mix is a 2X premixed formulation that can effectively reduce nonspecific priming and primer-dimer formation during PCR.
Form Solution
Format Solution
Quantity 100 Reactions
For Use With (Application) Standard PCR
No. of Reactions 100
Volume 50μL